The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163664.1 | NP_001157136.1 | 17 - 18 | 849 | 74 |
NM_001163666.1 | NP_001157138.1 | 15 - 16 | 822 | 74 | |
NM_001163667.1 | NP_001157139.1 | 16 - 17 | 837 | 74 | |
NM_001163668.1 | NP_001157140.1 | 15 - 16 | 798 | 74 | |
NM_001163669.1 | NP_001157141.1 | 16 - 17 | 810 | 74 | |
NM_001163670.1 | NP_001157142.1 | 13 - 14 | 765 | 74 | |
NM_011620.3 | NP_035750.2 | 14 - 15 | 777 | 74 | |
GenBank mRNA | AI322331.1 | - | 3 - 4 | 151 | 74 |
AK003147.1 | - | 13 - 14 | 732 | 74 | |
CK791812.1 | - | 6 - 7 | 487 | 74 | |
L48993.1 | - | 13 - 14 | 701 | 74 | |
L49018.1 | - | 11 - 12 | 695 | 74 | |
L49466.1 | - | 16 - 17 | 837 | 74 | |
L49467.1 | - | 17 - 18 | 849 | 74 | |
L49468.1 | - | 15 - 16 | 822 | 74 | |
L49470.1 | - | 16 - 17 | 810 | 74 | |
L49471.1 | - | 15 - 16 | 819 | 74 | |
L49472.1 | - | 15 - 16 | 798 | 74 | |
U77779.1 | - | 14 - 15 | 770 | 74 |