Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Top3b
Entrez Gene ID
21976
Gene Name
topoisomerase (DNA) III beta
Gene Aliases
-
Chromosome Location
Chr.16: 16870694 - 16892986 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011624.2 NP_035754.1 17 - 18 2392 99
GenBank mRNA AB013603.1 - 16 - 17 2205 99
AB045324.1 - 16 - 17 2942 99
AK144709.1 - 18 - 19 2378 99
AK146475.1 - 17 - 18 2414 99
AK163716.1 - 4 - 5 1254 99
AK163796.1 - 10 - 11 1401 99
BC031723.1 - 17 - 18 2392 99

View Full Product Details