Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sh3bp2
Entrez Gene ID
24055
Gene Name
SH3-domain binding protein 2
Gene Aliases
3BP2
Chromosome Location
Chr.5: 34525784 - 34563639 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145858.1 NP_001139330.1 6 - 7 732 76
NM_001145859.1 NP_001139331.1 6 - 7 735 76
NM_011893.3 NP_036023.3 6 - 7 632 76
GenBank mRNA AK089455.1 - 6 - 7 563 76
AK143548.1 - 6 - 7 737 76
AK150265.1 - 6 - 7 734 76
AK155898.1 - 6 - 7 632 76
BC085497.1 - 6 - 7 623 76
L14543.1 - 6 - 7 566 76

View Full Product Details