The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011894.2 | NP_036024.2 | 3 - 4 | 417 | 80 |
GenBank mRNA | AB016835.1 | - | 3 - 4 | 245 | 80 |
AK043375.1 | - | - | 462 | 80 | |
AK161427.1 | - | 2 - 3 | 399 | 80 | |
AK165033.1 | - | 3 - 4 | 451 | 80 | |
BC018237.1 | - | 3 - 4 | 449 | 80 | |
BC053741.1 | - | 3 - 4 | 417 | 80 |