The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199351.1 | NP_001186280.1 | 4 - 5 | 464 | 110 |
NM_001199352.1 | NP_001186281.1 | 4 - 5 | 328 | 110 | |
NM_012040.3 | NP_036170.1 | 4 - 5 | 404 | 110 | |
GenBank mRNA | AB023027.1 | - | 3 - 4 | 274 | 110 |
AF181984.1 | - | 4 - 5 | 378 | 110 | |
AK134665.1 | - | 4 - 5 | 335 | 110 | |
AK158755.1 | - | 4 - 5 | 329 | 110 | |
BC051996.1 | - | 4 - 5 | 311 | 110 | |
BC055891.1 | - | 4 - 5 | 304 | 110 | |
CX243772.1 | - | 4 - 5 | 464 | 110 |