Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pnck
Entrez Gene ID
93843
Gene Name
pregnancy upregulated non-ubiquitously expressed CaM kinase
Gene Aliases
Bstk3, CaMKIbeta2, CaMKlb2, Camk1b, Punc, caMKlb1
Chromosome Location
Chr.X: 73655992 - 73660127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199351.1 NP_001186280.1 7 - 8 806 72
NM_001199352.1 NP_001186281.1 7 - 8 670 72
NM_012040.3 NP_036170.1 7 - 8 746 72
GenBank mRNA AB023027.1 - 6 - 7 616 72
AF181984.1 - 7 - 8 720 72
AK134665.1 - 7 - 8 677 72
AK158755.1 - 7 - 8 671 72
BC051996.1 - 7 - 8 653 72
BC055891.1 - 7 - 8 646 72

View Full Product Details