Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prph
Entrez Gene ID
19132
Gene Name
peripherin
Gene Aliases
Prph1
Chromosome Location
Chr.15: 99053575 - 99058978 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163588.1 NP_001157060.1 1 - 2 586 119
NM_001163589.1 NP_001157061.1 1 - 2 586 119
NM_013639.2 NP_038667.2 1 - 2 586 119
GenBank mRNA AK149021.1 - 1 - 2 588 120
BC046291.1 - 1 - 2 579 119
X15475.1 - 1 - 2 348 119

View Full Product Details