The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252547.1 | NP_001239476.1 | 4 - 5 | 739 | 82 |
NM_013781.3 | NP_038809.1 | 5 - 6 | 1225 | 82 | |
NR_045535.1 | 6 - 7 | 1683 | 82 | ||
GenBank mRNA | AB030442.1 | - | 4 - 5 | 702 | 82 |
AB043953.1 | - | 5 - 6 | 1208 | 82 | |
AF168364.1 | - | 5 - 6 | 1166 | 82 | |
AK042709.1 | - | 4 - 5 | 723 | 82 | |
AK085885.1 | - | - | 723 | 80 | |
AK155165.1 | - | 5 - 6 | 1161 | 82 | |
AK172072.1 | - | 6 - 7 | 1604 | 82 | |
BC113203.1 | - | 5 - 6 | 1180 | 82 |