The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159630.1 | NP_001153102.1 | 7 - 8 | 841 | 62 |
NM_013883.2 | NP_038911.1 | 7 - 8 | 841 | 62 | |
GenBank mRNA | AB030906.1 | - | 7 - 8 | 740 | 62 |
AB221680.1 | - | 3 - 4 | 382 | 62 | |
AB221681.1 | - | 3 - 4 | 382 | 62 | |
BC034667.1 | - | 5 - 6 | 551 | 62 |