The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013918.2 | NP_038946.2 | 17 - 18 | 2222 | 91 |
GenBank mRNA | AF170563.1 | - | 17 - 18 | 2368 | 91 |
AK077769.1 | - | 2 - 3 | 395 | 91 | |
BC048171.1 | - | 17 - 18 | 2222 | 91 | |
BC063059.1 | - | 17 - 18 | 2222 | 91 |