Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nap1l1
Entrez Gene ID
53605
Gene Name
nucleosome assembly protein 1-like 1
Gene Aliases
AA407126, AI256722, D10Ertd68e, NAP-1
Chromosome Location
Chr.10: 111473192 - 111498150 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146707.1 NP_001140179.1 10 - 11 1618 71
NM_015781.4 NP_056596.1 11 - 12 1115 71
GenBank mRNA AK004633.1 - 11 - 12 1085 71
AK007322.1 - 11 - 12 1080 71
AK012250.1 - 7 - 8 626 71
AK032896.1 - 11 - 12 1088 71
AK050375.1 - 10 - 11 1620 71
AK136161.1 - 11 - 12 1081 71
AK145766.1 - 11 - 12 1079 71
AK152011.1 - 11 - 12 1009 71
AK153531.1 - 11 - 12 1009 71
AK169301.1 - 11 - 12 1027 71
AK216281.1 - 3 - 4 230 71
BC076591.1 - 10 - 11 1166 71
D12618.1 - 11 - 12 1071 71
X61449.1 - 8 - 9 788 71

View Full Product Details