The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190414.1 | NP_001177343.1 | 5 - 6 | 1333 | 96 |
NM_015828.3 | NP_056643.3 | 5 - 6 | 1151 | 96 | |
GenBank mRNA | AJ132236.1 | - | 4 - 5 | 979 | 96 |
AK010070.1 | - | 2 - 3 | 299 | 96 | |
AK136573.1 | - | 5 - 6 | 1152 | 96 | |
AK170707.1 | - | 5 - 6 | 1334 | 96 | |
AM697710.1 | - | 5 - 6 | 1072 | 96 | |
BC015277.1 | - | 5 - 6 | 1139 | 96 | |
BC051254.1 | - | 5 - 6 | 1139 | 96 |