The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016752.1 | NP_058032.1 | 6 - 7 | 745 | 59 |
GenBank mRNA | AK002961.1 | - | 6 - 7 | 739 | 59 |
AK007711.1 | - | 6 - 7 | 751 | 59 | |
AK160124.1 | - | 6 - 7 | 811 | 59 | |
BC002029.1 | - | 6 - 7 | 726 | 59 | |
D87990.1 | - | 6 - 7 | 745 | 59 |