Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35b1
Entrez Gene ID
110172
Gene Name
solute carrier family 35, member B1
Gene Aliases
Ugalt2
Chromosome Location
Chr.11: 95384891 - 95391776 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016752.1 NP_058032.1 8 - 9 1003 79
GenBank mRNA AK002961.1 - 8 - 9 997 79
AK007711.1 - 8 - 9 1009 79
AK160124.1 - 8 - 9 1070 79
AK213577.1 - 1 - 2 90 79
BC002029.1 - 8 - 9 984 79
D87990.1 - 8 - 9 1003 79

View Full Product Details