Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nucb2
Entrez Gene ID
53322
Gene Name
nucleobindin 2
Gene Aliases
AI607786, Calnuc, Nefa, Nesfatin-1
Chromosome Location
Chr.7: 116504353 - 116540588 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130479.2 NP_001123951.1 2 - 3 294 115
GenBank mRNA AJ222586.1 - 2 - 3 294 115
AK145934.1 - 2 - 3 249 115
AK160442.1 - 1 - 2 142 114
BC010459.1 - 2 - 3 208 115

View Full Product Details