Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nucb2
Entrez Gene ID
53322
Gene Name
nucleobindin 2
Gene Aliases
AI607786, Calnuc, Nefa, Nesfatin-1
Chromosome Location
Chr.7: 116504353 - 116540588 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130479.2 NP_001123951.1 4 - 5 543 92
GenBank mRNA AJ222586.1 - 4 - 5 543 92
AK145934.1 - 4 - 5 498 92
AK160442.1 - 3 - 4 391 92
BC010459.1 - 4 - 5 457 92

View Full Product Details