The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016914.2 | NP_058610.1 | 4 - 5 | 665 | 83 |
GenBank mRNA | AF202533.1 | - | 4 - 5 | 665 | 83 |
AK158110.1 | - | 4 - 5 | 685 | 83 | |
BC104388.1 | - | 3 - 4 | 575 | 83 | |
BC104389.1 | - | 2 - 3 | 268 | 83 |