Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Akap8l
Entrez Gene ID
54194
Gene Name
A kinase (PRKA) anchor protein 8-like
Gene Aliases
HAP95, Nakap95
Chromosome Location
Chr.17: 32321424 - 32350623 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017476.2 NP_059504.2 5 - 6 894 72
GenBank mRNA AB028921.1 - 5 - 6 887 72
AK011325.1 - 5 - 6 899 72
AK166526.1 - 6 - 7 810 72
BC046801.1 - 5 - 6 883 72
BC051015.1 - 5 - 6 894 72

View Full Product Details