The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017476.2 | NP_059504.2 | 12 - 13 | 1610 | 87 |
GenBank mRNA | AB028921.1 | - | 12 - 13 | 1606 | 87 |
AK011325.1 | - | 12 - 13 | 1615 | 87 | |
AK166526.1 | - | 13 - 14 | 1526 | 87 | |
BC003242.1 | - | 2 - 3 | 239 | 87 | |
BC046801.1 | - | 12 - 13 | 1599 | 87 | |
BC051015.1 | - | 10 - 11 | 3919 | 87 |