Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trpm1
Entrez Gene ID
17364
Gene Name
transient receptor potential cation channel, subfamily M, member 1
Gene Aliases
4732499L03Rik, AI606771, Ltrpc1, Mlsn1
Chromosome Location
Chr.7: 64153835 - 64269772 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039104.2 NP_001034193.2 4 - 5 760 108
NM_018752.3 NP_061222.3 4 - 5 653 108
GenBank mRNA AF047714.1 - 4 - 5 522 108
AK044507.1 - 4 - 5 751 108
AK148094.1 - 4 - 5 558 108
AK148193.1 - 4 - 5 561 108
AK148268.1 - 4 - 5 507 108
AK148275.1 - 4 - 5 563 108
AK149290.1 - 4 - 5 682 108
AK164395.1 - 4 - 5 563 108
AK164423.1 - 4 - 5 570 108
AK196403.1 - 2 - 3 335 108
AK205201.1 - 1 - 2 211 108
AK212724.1 - 1 - 2 180 108
AK219645.1 - 1 - 2 226 108
AY180104.2 - 4 - 5 760 108
BC082560.1 - 4 - 5 529 108
BC085168.1 - 3 - 4 332 108

View Full Product Details