Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stx8
Entrez Gene ID
55943
Gene Name
syntaxin 8
Gene Aliases
0610007H08Rik, 1110002H11Rik, 4930571E13Rik, CARB
Chromosome Location
Chr.11: 67966411 - 68207148 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018768.2 NP_061238.1 4 - 5 335 77
GenBank mRNA AB040054.1 - 4 - 5 332 77
AF036716.1 - 4 - 5 322 77
AF093064.1 - 4 - 5 344 77
AK003297.1 - 4 - 5 361 77
BC038186.1 - 4 - 5 334 77
BC048167.1 - 4 - 5 335 77
BC048479.1 - 4 - 5 360 77

View Full Product Details