The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018783.4 | NP_061253.2 | 3 - 4 | 509 | 59 |
GenBank mRNA | AF290474.3 | - | 3 - 4 | 509 | 59 |
AK147973.1 | - | 3 - 4 | 460 | 59 | |
AK148182.1 | - | 3 - 4 | 515 | 59 | |
AK148260.1 | - | 3 - 4 | 440 | 59 | |
AK161154.1 | - | 3 - 4 | 513 | 59 | |
AK164414.1 | - | 3 - 4 | 461 | 59 | |
AK217890.1 | - | 1 - 2 | 36 | 59 | |
BC017682.1 | - | 3 - 4 | 444 | 59 |