The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165949.1 | NP_001159421.1 | 2 - 3 | 541 | 107 |
NM_018812.2 | NP_061282.2 | 3 - 4 | 436 | 107 | |
NM_146135.2 | NP_666247.1 | 2 - 3 | 578 | 107 | |
GenBank mRNA | AF034080.3 | - | 2 - 3 | 313 | 107 |
AK040619.1 | - | 2 - 3 | 569 | 107 | |
AK077551.1 | - | 2 - 3 | 527 | 107 | |
AK085503.1 | - | 2 - 3 | 512 | 107 | |
AK211740.1 | - | 2 - 3 | 142 | 107 | |
BC023128.1 | - | 2 - 3 | 484 | 107 | |
BC050988.1 | - | 3 - 4 | 395 | 107 | |
BC051252.1 | - | 2 - 3 | 451 | 107 |