The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290674.1 | NP_001277603.1 | 27 - 27 | 6508 | 74 |
NM_001290675.1 | NP_001277604.1 | 27 - 27 | 6475 | 74 | |
GenBank mRNA | AK220540.1 | - | 1 - 1 | 733 | 74 |
BC158048.1 | - | 27 - 27 | 6166 | 74 | |
BC172147.1 | - | 27 - 27 | 6199 | 74 | |
L42338.1 | - | 1 - 1 | 947 | 74 |