The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019417.3 | NP_062290.2 | 1 - 2 | 180 | 66 |
GenBank mRNA | AB221672.1 | - | 1 - 2 | 95 | 66 |
AK035513.1 | - | 1 - 2 | 155 | 66 | |
BC030068.1 | - | 1 - 2 | 109 | 66 | |
Y08361.1 | - | 1 - 2 | 131 | 66 |