The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019501.3 | NP_062374.2 | 9 - 10 | 1117 | 76 |
GenBank mRNA | AB210841.1 | - | 9 - 10 | 1037 | 76 |
AF118855.1 | - | 7 - 8 | 812 | 76 | |
BC026820.1 | - | 9 - 10 | 1019 | 76 | |
BC107273.1 | - | 9 - 10 | 932 | 76 | |
BC107274.1 | - | 9 - 10 | 936 | 76 |