The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019662.2 | NP_062636.2 | 4 - 5 | 911 | 94 |
GenBank mRNA | AF084466.1 | - | 3 - 4 | 654 | 94 |
AK077271.1 | - | 4 - 5 | 909 | 94 | |
AK087473.1 | - | 4 - 5 | 884 | 94 | |
AK159550.1 | - | 3 - 4 | 694 | 94 | |
BC057138.1 | - | 4 - 5 | 890 | 94 |