The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033988.2 | NP_001029160.1 | 7 - 8 | 854 | 73 |
NM_001284319.1 | NP_001271248.1 | 7 - 8 | 846 | 73 | |
NM_019744.4 | NP_062718.2 | 7 - 8 | 764 | 73 | |
GenBank mRNA | AF159461.1 | - | 7 - 8 | 761 | 73 |
AK014479.1 | - | 7 - 8 | 762 | 73 | |
AK032951.1 | - | 5 - 6 | 520 | 73 | |
AK129020.1 | - | 7 - 8 | 812 | 73 | |
AK132981.1 | - | - | 73 | ||
AK145070.1 | - | 7 - 8 | 815 | 73 | |
AK145945.1 | - | 7 - 8 | 807 | 73 | |
AK145954.1 | - | 7 - 8 | 811 | 73 | |
AK146384.1 | - | 7 - 8 | 765 | 73 | |
AK146498.1 | - | 7 - 8 | 804 | 73 | |
AK146598.1 | - | 7 - 8 | 788 | 73 | |
AK146646.1 | - | 7 - 8 | 759 | 73 | |
AK146649.1 | - | 7 - 8 | 761 | 73 | |
AK147036.1 | - | 7 - 8 | 762 | 73 | |
AK147125.1 | - | 7 - 8 | 806 | 73 | |
AK156999.1 | - | 7 - 8 | 846 | 73 | |
BC031528.1 | - | 7 - 8 | 805 | 73 | |
BC085086.1 | - | 7 - 8 | 750 | 73 |