The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019754.3 | NP_062728.1 | 2 - 3 | 586 | 103 |
GenBank mRNA | AB031291.1 | - | 2 - 3 | 419 | 103 |
AK013484.1 | - | 2 - 3 | 490 | 103 | |
BC043027.1 | - | 2 - 3 | 365 | 103 | |
BC055338.1 | - | 2 - 3 | 586 | 103 |