Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc5a1
Entrez Gene ID
20537
Gene Name
solute carrier family 5 (sodium/glucose cotransporter), member 1
Gene Aliases
Sglt1
Chromosome Location
Chr.5: 33104219 - 33162699 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019810.4 NP_062784.3 12 - 13 1685 65
GenBank mRNA AF163846.1 - 12 - 13 1447 65
AF208031.1 - 12 - 13 1685 65
AK033512.1 - 12 - 13 1671 65
AK085602.1 - 12 - 13 1476 65
BC003845.1 - 12 - 13 1665 65

View Full Product Details