Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtm1
Entrez Gene ID
17772
Gene Name
X-linked myotubular myopathy gene 1
Gene Aliases
AF073996, Mtm, mKIAA4176
Chromosome Location
Chr.X: 71210767 - 71315413 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164190.1 NP_001157662.1 2 - 3 258 105
NM_001164191.1 NP_001157663.1 2 - 3 149 105
NM_001164192.1 NP_001157664.1 2 - 3 149 105
NM_001164193.1 NP_001157665.1 2 - 3 149 105
NM_019926.3 NP_064310.2 2 - 3 138 105
GenBank mRNA AF073996.1 - 2 - 3 139 105
AK016445.1 - 2 - 3 259 105
AK031794.1 - 2 - 3 120 105
AK032870.1 - 2 - 3 102 105
AK149997.1 - 2 - 3 151 105
AK166759.1 - 2 - 3 136 105
AK171475.1 - 2 - 3 127 105
BC051022.1 - 2 - 3 107 105
BC090984.1 - 2 - 3 116 105

View Full Product Details