Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf14
Entrez Gene ID
56736
Gene Name
ring finger protein 14
Gene Aliases
2310075C09Rik, 2610005D23Rik, AA986456, AU041447, D18Ertd188e, D7Bwg0165e, Triad2
Chromosome Location
Chr.18: 38296635 - 38317852 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164621.1 NP_001158093.1 8 - 9 1895 98
NM_001164622.1 NP_001158094.1 6 - 7 1430 98
NM_020012.2 NP_064396.2 7 - 8 1582 98
GenBank mRNA AF249667.1 - 7 - 8 1528 98
AF249668.1 - 5 - 6 1113 98
AK009688.1 - 2 - 3 193 98
AK011316.2 - 8 - 9 1583 98
AK038986.1 - 6 - 7 1372 98
BC049079.1 - 8 - 9 1895 98
BC052205.1 - - 3591 98
BC054841.1 - 7 - 8 1512 98
BC094250.1 - 7 - 8 1533 98

View Full Product Details