Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc6a14
Entrez Gene ID
56774
Gene Name
solute carrier family 6 (neurotransmitter transporter), member 14
Gene Aliases
1110007A17Rik, 9030613J17Rik, ATB0plus, CATB0plus
Chromosome Location
Chr.X: 21714900 - 21742358 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020049.4 NP_064433.3 9 - 10 1516 93
GenBank mRNA AB033285.1 - 9 - 10 1274 93
AF161714.1 - 9 - 10 1516 93
AF320226.1 - 9 - 10 1460 93
AK018553.1 - 9 - 10 1375 93
BC129908.1 - 9 - 10 1307 93
BC129909.1 - 9 - 10 1307 93

View Full Product Details