The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205085.1 | NP_001192014.1 | 1 - 2 | 587 | 63 |
NM_020496.3 | NP_065242.1 | 1 - 2 | 587 | 63 | |
NM_194263.2 | NP_919239.1 | 1 - 2 | 587 | 63 | |
GenBank mRNA | AF306667.1 | - | 1 - 2 | 525 | 63 |
AJ277486.1 | - | 1 - 2 | 129 | 63 | |
AK052155.1 | - | 1 - 2 | 569 | 63 | |
AK052389.1 | - | 1 - 2 | 570 | 63 | |
AK141251.1 | - | 1 - 2 | 357 | 63 |