The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285487.1 | NP_001272416.1 | 10 - 11 | 896 | 79 |
NM_001285488.1 | NP_001272417.1 | 9 - 10 | 790 | 79 | |
NM_021461.5 | NP_067436.1 | 8 - 9 | 746 | 79 | |
GenBank mRNA | AK010697.1 | - | 8 - 9 | 724 | 79 |
AK141298.1 | - | 8 - 9 | 724 | 79 | |
AK155934.1 | - | 10 - 11 | 857 | 79 | |
AK169982.1 | - | 9 - 10 | 756 | 79 | |
BC021369.1 | - | 8 - 9 | 702 | 79 | |
BQ551007.1 | - | 6 - 7 | 453 | 79 | |
Y11091.1 | - | 8 - 9 | 711 | 79 |