The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021526.2 | NP_067501.2 | 10 - 11 | 1077 | 101 |
GenBank mRNA | AK012013.1 | - | 10 - 11 | 1008 | 101 |
AK031863.1 | - | 10 - 11 | 1014 | 101 | |
AK078134.1 | - | 10 - 11 | 1029 | 101 | |
AK150280.1 | - | 10 - 11 | 1013 | 101 | |
AK151104.1 | - | 10 - 11 | 1060 | 101 | |
AK151733.1 | - | 10 - 11 | 1060 | 101 | |
BC003742.1 | - | 10 - 11 | 982 | 101 | |
Y13071.1 | - | 10 - 11 | 998 | 101 |