The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127351.1 | NP_001120823.1 | 3 - 4 | 526 | 75 |
NM_001177804.1 | NP_001171275.1 | 3 - 4 | 518 | 75 | |
NM_022433.2 | NP_071878.2 | 3 - 4 | 571 | 75 | |
GenBank mRNA | AF299338.1 | - | 3 - 4 | 571 | 75 |
AF299339.1 | - | 3 - 4 | 526 | 75 | |
AK075861.1 | - | 3 - 4 | 500 | 75 | |
BC025878.1 | - | 3 - 4 | 470 | 75 | |
CB953804.1 | - | 3 - 4 | 518 | 75 | |
CT010402.1 | - | 2 - 3 | 277 | 75 | |
EU886466.1 | - | 3 - 4 | 516 | 75 | |
FJ621493.1 | - | 3 - 4 | 514 | 75 |