The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168515.1 | NP_001161987.1 | 2 - 3 | 169 | 76 |
NM_023879.3 | NP_076368.1 | 2 - 3 | 169 | 76 | |
GenBank mRNA | AK020800.1 | - | 2 - 3 | 151 | 76 |
AY008297.1 | - | 2 - 3 | 151 | 76 | |
AY914560.1 | - | 2 - 3 | 145 | 76 | |
BC016092.1 | - | 2 - 3 | 131 | 76 | |
BC081545.1 | - | 2 - 3 | 169 | 76 | |
BC086692.1 | - | 2 - 3 | 133 | 76 | |
HM067441.1 | - | 1 - 2 | 73 | 76 |