The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025590.4 | NP_079866.2 | 8 - 9 | 1002 | 66 |
GenBank mRNA | AF416923.1 | - | 8 - 9 | 940 | 66 |
AK031588.1 | - | 8 - 9 | 919 | 66 | |
AK038546.1 | - | 8 - 9 | 940 | 66 | |
AK048670.1 | - | 8 - 9 | 1002 | 66 | |
AK052332.1 | - | 8 - 9 | 1018 | 66 | |
BC042492.1 | - | 8 - 9 | 937 | 66 |