The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026385.4 | NP_080661.1 | 1 - 2 | 208 | 91 |
GenBank mRNA | AK002477.1 | - | 1 - 2 | 209 | 91 |
AK018198.1 | - | 1 - 2 | 198 | 90 | |
AK032540.1 | - | 1 - 2 | 197 | 91 | |
BC024534.1 | - | 1 - 2 | 199 | 91 |