Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gar1
Entrez Gene ID
68147
Gene Name
GAR1 ribonucleoprotein homolog (yeast)
Gene Aliases
AA409823, AI326794, C430047J18Rik, Nola1
Chromosome Location
Chr.3: 129824912 - 129831396 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026578.3 NP_080854.1 1 - 2 281 54
GenBank mRNA AK021256.1 - 1 - 2 42 54
BC021873.1 - 1 - 2 187 54
BC048685.1 - 1 - 2 206 54

View Full Product Details