Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Robo4
Entrez Gene ID
74144
Gene Name
roundabout homolog 4 (Drosophila)
Gene Aliases
1200012D01Rik, AI593217
Chromosome Location
Chr.9: 37401902 - 37414023 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF536772.1 - 17 - 18 3068 72
AK004723.2 - 17 - 18 3044 72
AK030973.1 - - 1835 72
AK087355.1 - 16 - 17 3220 72
AK172104.1 - 16 - 17 3306 72
BC020129.1 - 9 - 10 1588 72
BC071193.1 - 16 - 17 2758 72

View Full Product Details