The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024954.1 | NP_001020125.1 | 7 - 8 | 1052 | 118 |
GenBank mRNA | AJ300184.1 | - | 4 - 5 | 486 | 118 |
AK010506.1 | - | 4 - 5 | 531 | 118 | |
AK038786.1 | - | - | 656 | 118 | |
AK076968.1 | - | 5 - 6 | 677 | 118 | |
BC032875.1 | - | 6 - 7 | 748 | 118 |