Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mynn
Entrez Gene ID
80732
Gene Name
myoneurin
Gene Aliases
2810011C24Rik, AA415053, AI661031, AW049392, Mynr, SBBIZ1
Chromosome Location
Chr.3: 30602087 - 30619873 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289621.1 NP_001276550.1 3 - 4 1694 94
NM_001289622.1 NP_001276551.1 3 - 4 1564 94
NM_001289623.1 NP_001276552.1 3 - 4 1564 94
NM_030557.3 NP_085034.2 3 - 4 1694 94
GenBank mRNA AF349561.1 - 2 - 3 1086 94
AK014315.1 - 3 - 4 1145 94
AK019238.2 - 3 - 4 1138 94
AK029657.1 - 3 - 4 985 94
AK030898.1 - 3 - 4 1020 94
AK136107.1 - 3 - 4 1143 94
AK147959.1 - 3 - 4 1018 94
AK163261.1 - 3 - 4 1008 94
AK163264.1 - 3 - 4 1008 94
BC007477.1 - 3 - 4 1007 94
BC065084.1 - 3 - 4 1694 94

View Full Product Details