Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekha3
Entrez Gene ID
83435
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 3
Gene Aliases
FAPP1
Chromosome Location
Chr.2: 76675310 - 76697335 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031256.3 NP_112546.1 3 - 4 589 100
GenBank mRNA AF286163.1 - 3 - 4 378 100
BC012283.1 - 3 - 4 329 100
BC031110.1 - 4 - 5 397 100

View Full Product Details