Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tktl1
Entrez Gene ID
83553
Gene Name
transketolase-like 1
Gene Aliases
C79967
Chromosome Location
Chr.X: 74177259 - 74208498 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031379.2 NP_113556.2 12 - 13 1651 69
GenBank mRNA AF285571.1 - 12 - 13 1626 69
AJ627042.1 - 5 - 6 541 69
AK029546.1 - 12 - 13 1652 69
AK139841.1 - 12 - 13 1651 69
BC113768.1 - 12 - 13 1626 69

View Full Product Details