The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032465.2 | NP_115854.2 | 6 - 7 | 1043 | 98 |
GenBank mRNA | AK040329.1 | - | 6 - 7 | 930 | 98 |
AK040539.1 | - | 6 - 7 | 1047 | 98 | |
AK154076.1 | - | 6 - 7 | 1013 | 98 | |
AK156470.1 | - | 6 - 7 | 1044 | 98 | |
AY029156.1 | - | 6 - 7 | 904 | 98 | |
BC052865.1 | - | 6 - 7 | 940 | 98 |