The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133215.1 | NP_573478.1 | 3 - 4 | 165 | 63 |
GenBank mRNA | AF262986.1 | - | 3 - 4 | 165 | 63 |
AK146641.1 | - | 3 - 4 | 182 | 63 |