Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn1r78
Entrez Gene ID
171242
Gene Name
vomeronasal 1 receptor 78
Gene Aliases
V1rg7
Chromosome Location
Chr.7: 12152464 - 12153405 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134208.2 NP_598969.2 1 - 1 859 144
GenBank mRNA BC107195.1 - 1 - 1 859 144

View Full Product Details