The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289709.1 | NP_001276638.1 | 11 - 12 | 1204 | 65 |
NM_001289710.1 | NP_001276639.1 | 9 - 10 | 1126 | 65 | |
NM_139302.2 | NP_647463.1 | 10 - 11 | 1189 | 65 | |
GenBank mRNA | AF426314.1 | - | 10 - 11 | 1086 | 65 |
AK129455.1 | - | 9 - 10 | 2567 | 65 | |
AK152409.1 | - | 10 - 11 | 1119 | 65 | |
AK152502.1 | - | 9 - 10 | 1077 | 65 | |
AK155248.1 | - | 11 - 12 | 1129 | 65 | |
AK167762.1 | - | 10 - 11 | 1162 | 65 | |
BC024477.1 | - | 10 - 11 | 1104 | 65 | |
BC028859.1 | - | 8 - 9 | 1528 | 65 | |
EF676083.1 | - | 10 - 11 | 1165 | 65 |